Assay Details
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005667339.3 | XP_005667396.1 | 500 | 115 | ||
XM_021062791.1 | XP_020918450.1 | 431 | 115 | |||
GenBank mRNA | AK233932.1 | - | 5 - 6 | 366 | 115 | |
AY609879.1 | - | 5 - 6 | 360 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
