Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126434.6 | XP_003126482.3 | 411 | 89 | ||
| GenBank mRNA | AK234095.1 | - | 1 - 2 | 364 | 89 | |
| AK351904.1 | - | 2 - 3 | 361 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003126434.6 transcripts.
Set Membership: |
|

KCNJ8
Genomic Map