Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126434.6 | XP_003126482.3 | 837 | 90 | ||
| GenBank mRNA | AK234095.1 | - | 2 - 3 | 790 | 90 | |
| AK351904.1 | - | 3 - 4 | 787 | 90 | ||
| EU655629.1 | - | 1 - 2 | 80 | 90 | ||
More Information
Set Membership: |
|

KCNJ8
Genomic Map