Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356734.4 | XP_003356782.2 | 1811 | 110 | ||
| GenBank mRNA | AK234436.1 | - | 1 | 323 | 110 | |
| AK392445.1 | - | 1 | 1506 | 110 | ||
| AK400715.1 | - | 1 | 1560 | 110 | ||
More Information
Set Membership: |
|

HSPA2
Genomic Map