Assay Details
Gene Symbol: SRPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924519.5 | XP_001924554.3 | 1711 | 57 | ||
GenBank mRNA | AK234573.1 | - | 10 - 11 | 1708 | 57 | |
AK347291.1 | - | 10 - 11 | 1682 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
