Assay Details
| Gene Symbol: KCNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928032.7 | XP_001928067.1 | 752 | 98 | ||
| GenBank mRNA | AK235536.1 | - | 1 - 2 | 421 | 98 | |
| AK389883.1 | - | 1 - 2 | 706 | 98 | ||
| AK390480.1 | - | 1 - 2 | 482 | 98 | ||
More Information
Set Membership: |
|

KCNK1
Genomic Map