Assay Details
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095008.1 | XP_020950667.1 | 437 | 98 | ||
| GenBank mRNA | AB198667.1 | - | 2 - 3 | 267 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095008.1 | XP_020950667.1 | 437 | 98 | ||
| GenBank mRNA | AB198667.1 | - | 2 - 3 | 267 | 98 | |
Set Membership: |
|