Assay Details
| Gene Symbol: MYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986326.2 | XP_013841780.1 | 1207 | 75 | ||
| GenBank mRNA | AB527070.1 | - | 6 - 7 | 961 | 75 | |
| AK240259.1 | - | 6 - 7 | 971 | 75 | ||
| AK346809.1 | - | 6 - 7 | 975 | 75 | ||
| AK351021.1 | - | 6 - 7 | 959 | 75 | ||
| AK400734.1 | - | 6 - 7 | 989 | 75 | ||
More Information
Set Membership: |
|

MYG1
Genomic Map