Assay Details
| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925047.5 | XP_001925082.1 | 631 | 69 | ||
| GenBank mRNA | AK230691.1 | - | 2 - 3 | 220 | 69 | |
More Information
Set Membership: |
|

SERPINB7
Genomic Map