Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981660.2 | XP_013837114.2 | 475 | 76 | ||
| XM_021070630.1 | XP_020926289.1 | 1284 | 76 | |||
| XM_021070631.1 | XP_020926290.1 | 719 | 76 | |||
| XM_021070632.1 | XP_020926291.1 | 632 | 76 | |||
| XM_021070633.1 | XP_020926292.1 | 699 | 76 | |||
| GenBank mRNA | AK235996.1 | - | 3 - 4 | 410 | 76 | |
| AK350412.1 | - | 5 - 6 | 660 | 76 | ||
| AK351313.1 | - | 4 - 5 | 603 | 76 | ||
| AK393537.1 | - | 5 - 6 | 739 | 76 | ||
| AK394949.1 | - | 3 - 4 | 423 | 76 | ||
| AK396632.1 | - | 3 - 4 | 402 | 76 | ||
| AK400811.1 | - | 3 - 4 | 412 | 76 | ||
| AK400958.1 | - | 3 - 4 | 434 | 76 | ||
More Information
Set Membership: |
|

SHISA5
Genomic Map