Assay Details
Gene Symbol: CNOT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124884.6 | XP_003124932.2 | 757 | 84 | ||
GenBank mRNA | AK236147.1 | - | 2 - 3 | 295 | 84 | |
AK350398.1 | - | 2 - 3 | 176 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
