Assay Details
Gene Symbol: C1QBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067745.1 | XP_020923404.1 | 581 | 57 | ||
GenBank mRNA | AK236153.1 | - | 3 - 4 | 558 | 57 | |
AK394353.1 | - | 3 - 4 | 527 | 57 | ||
AK399044.1 | - | 3 - 4 | 527 | 57 | ||
LN835347.1 | - | 3 - 4 | 476 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
