Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657925.3 | XP_005657982.1 | 2128 | 77 | ||
| XM_013991020.2 | XP_013846474.1 | 3535 | 77 | |||
| XM_021080051.1 | XP_020935710.1 | 3616 | 77 | |||
| XM_021080052.1 | XP_020935711.1 | 3613 | 77 | |||
| XM_021080053.1 | XP_020935712.1 | 2468 | 77 | |||
| XM_021080055.1 | XP_020935714.1 | 3325 | 77 | |||
| XM_021080056.1 | XP_020935715.1 | 3244 | 77 | |||
| XM_021080057.1 | XP_020935716.1 | 3616 | 77 | |||
| XM_021080058.1 | XP_020935717.1 | 3616 | 77 | |||
| GenBank mRNA | AK236209.1 | - | 1 - 2 | 119 | 77 | |
More Information
Set Membership: |
|

MAP7D3
Genomic Map