Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123198.4 | XP_003123246.1 | 835 | 71 | ||
| GenBank mRNA | AK236367.1 | - | 1 - 2 | 213 | 71 | |
| AK397933.1 | - | 1 - 2 | 216 | 71 | ||
More Information
Set Membership: |
|

SNAPC2
Genomic Map