Assay Details
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131552.4 | XP_003131600.1 | 548 | 63 | ||
GenBank mRNA | AK236888.1 | - | 4 - 5 | 462 | 63 | |
AK240159.1 | - | 4 - 5 | 477 | 63 | ||
AK392793.1 | - | 4 - 5 | 482 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
