Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358401.5 | XP_003358449.3 | 2065 | 94 | ||
| XM_013981553.1 | XP_013837007.1 | 1959 | 94 | |||
| GenBank mRNA | AK237191.1 | - | 1 | 921 | 94 | |
More Information
Set Membership: |
|

ZDHHC3
Genomic Map