Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123993.6 | XP_003124041.5 | 655 | 70 | ||
| GenBank mRNA | AK237276.1 | - | 4 - 5 | 634 | 70 | |
| AK345732.1 | - | 4 - 5 | 589 | 70 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map