Assay Details
Gene Symbol: DDX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928544.4 | XP_001928579.1 | 1879 | 120 | ||
XM_021090025.1 | XP_020945684.1 | 1995 | 120 | |||
XM_021090026.1 | XP_020945685.1 | 2019 | 120 | |||
GenBank mRNA | AK237768.1 | - | 1 | 120 | 120 | |
AK344951.1 | - | 11 | 1794 | 120 | ||
AK389316.1 | - | 1 | 228 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
