Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131737.4 | XP_003131785.1 | 710 | 83 | ||
| XM_005669064.3 | XP_005669121.1 | 710 | 83 | |||
| GenBank mRNA | AK231104.1 | - | 3 - 4 | 643 | 83 | |
More Information
Set Membership: |
|

TEFM
Genomic Map