Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133320.4 | XP_003133368.1 | 306 | 94 | ||
| GenBank mRNA | AK231270.1 | - | 1 - 2 | 211 | 94 | |
| AK400393.1 | - | 1 - 2 | 245 | 94 | ||
More Information
Set Membership: |
|

SPCS3
Genomic Map