Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130656.3 | XP_003130704.1 | 358 | 102 | ||
| XM_005668049.2 | XP_005668106.1 | 358 | 102 | |||
| GenBank mRNA | AK231493.1 | - | 3 - 4 | 298 | 102 | |
| AK346601.1 | - | 3 - 4 | 303 | 102 | ||
| AK351267.1 | - | 3 - 4 | 302 | 102 | ||
| AK391979.1 | - | 3 - 4 | 296 | 102 | ||
| AK399829.1 | - | 3 - 4 | 297 | 102 | ||
| AY610334.1 | - | 3 - 4 | 269 | 102 | ||
More Information
Set Membership: |
|

DCTN3
Genomic Map