Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013982786.2 | XP_013838240.1 | 997 | 55 | ||
| XM_021072266.1 | XP_020927925.1 | 847 | 55 | |||
| GenBank mRNA | AK231498.1 | - | 2 - 3 | 551 | 55 | |
| AK237639.1 | - | 2 - 3 | 536 | 55 | ||
| AK240404.1 | - | 2 - 3 | 549 | 55 | ||
| AK390034.1 | - | 2 - 3 | 551 | 55 | ||
More Information
Set Membership: |
|

RILPL2
Genomic Map