Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098870.1 | XP_020954529.1 | 524 | 81 | ||
| GenBank mRNA | AK238960.1 | - | 5 - 6 | 479 | 81 | |
| AK348895.1 | - | 5 - 6 | 481 | 81 | ||
More Information
Set Membership: |
|

EFL1
Genomic Map