Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359868.5 | XP_003359916.2 | 82 | |||
| GenBank mRNA | AK239094.1 | - | 1 | 423 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359868.5 | XP_003359916.2 | 82 | |||
| GenBank mRNA | AK239094.1 | - | 1 | 423 | 82 | |
Set Membership: |
|