Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124436.4 | XP_003124484.1 | 1268 | 89 | ||
| XM_003481011.3 | XP_003481059.1 | 1103 | 89 | |||
| XM_021086358.1 | XP_020942017.1 | 1018 | 89 | |||
| GenBank mRNA | AK239162.1 | - | 6 - 7 | 1200 | 89 | |
More Information
Set Membership: |
|

TMEM248
Genomic Map