Assay Details
| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129723.4 | XP_003129771.1 | 3570 | 64 | ||
| GenBank mRNA | AK240419.1 | - | 1 | 1115 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129723.4 | XP_003129771.1 | 3570 | 64 | ||
| GenBank mRNA | AK240419.1 | - | 1 | 1115 | 64 | |
Set Membership: |
|