Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359315.4 | XP_003359363.1 | 719 | 111 | ||
| GenBank mRNA | AK240543.1 | - | 1 - 2 | 266 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003359315.4 transcripts.
Set Membership: |
|

UBTD1
Genomic Map