Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359315.4 | XP_003359363.1 | 950 | 60 | ||
| XM_005657441.3 | XP_005657498.1 | 1070 | 60 | |||
| XM_013990469.2 | XP_013845923.1 | 2256 | 60 | |||
| XM_013990470.2 | XP_013845924.1 | 1059 | 60 | |||
| XM_021074394.1 | XP_020930053.1 | 882 | 60 | |||
| GenBank mRNA | AK240543.1 | - | 2 - 3 | 497 | 60 | |
More Information
Set Membership: |
|

UBTD1
Genomic Map