Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101553.1 | XP_020957212.1 | 1251 | 78 | ||
| GenBank mRNA | AK343688.1 | - | 7 - 8 | 695 | 78 | |
| AK390166.1 | - | 7 - 8 | 703 | 78 | ||
More Information
Set Membership: |
|

ZNF330
Genomic Map