Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094221.1 | XP_020949880.1 | 413 | 80 | ||
| GenBank mRNA | AK344568.1 | - | 4 - 5 | 316 | 80 | |
More Information
Set Membership: |
|

CNEP1R1
Genomic Map