Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126376.6 | XP_003126424.3 | 1192 | 75 | ||
| GenBank mRNA | AK344682.1 | - | 4 - 5 | 892 | 75 | |
More Information
Set Membership: |
|

TMEM19
Genomic Map