Assay Details
| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064906.1 | XP_020920565.1 | 7429 | 98 | ||
| XM_021064907.1 | XP_020920566.1 | 7062 | 98 | |||
| GenBank mRNA | AK344722.1 | - | 1 | 121 | 98 | |
More Information
Set Membership: |
|

GPR158
Genomic Map