Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092557.1 | XP_020948216.1 | 8291 | 98 | ||
| XM_021092558.1 | XP_020948217.1 | 8288 | 98 | |||
| XM_021092559.1 | XP_020948218.1 | 4178 | 98 | |||
| XM_021092560.1 | XP_020948219.1 | 4175 | 98 | |||
| XM_021092562.1 | XP_020948221.1 | 4256 | 98 | |||
| XM_021092563.1 | XP_020948222.1 | 4045 | 98 | |||
| XM_021092564.1 | XP_020948223.1 | 4222 | 98 | |||
| GenBank mRNA | AK345181.1 | - | 1 | 1323 | 98 | |
More Information
Set Membership: |
|

PRICKLE1
Genomic Map