Assay Details
| Gene Symbol: JADE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123948.5 | XP_003123996.3 | 6407 | 75 | ||
| XM_005655015.2 | XP_005655072.1 | 8498 | 75 | |||
| XM_013987596.2 | XP_013843050.1 | 7148 | 75 | |||
| XM_013987597.2 | XP_013843051.1 | 6590 | 75 | |||
| XM_021084767.1 | XP_020940426.1 | 6649 | 75 | |||
| XM_021084768.1 | XP_020940427.1 | 8023 | 75 | |||
| XM_021084769.1 | XP_020940428.1 | 6586 | 75 | |||
| XM_021084770.1 | XP_020940429.1 | 6397 | 75 | |||
| XM_021084771.1 | XP_020940430.1 | 8678 | 75 | |||
| XM_021084772.1 | XP_020940431.1 | 7839 | 75 | |||
| XM_021084773.1 | XP_020940432.1 | 7394 | 75 | |||
| XM_021084774.1 | XP_020940433.1 | 8369 | 75 | |||
| XM_021084775.1 | XP_020940434.1 | 6452 | 75 | |||
| XM_021084776.1 | XP_020940435.1 | 7895 | 75 | |||
| XM_021084777.1 | XP_020940436.1 | 6275 | 75 | |||
| GenBank mRNA | AK346143.1 | - | 1 | 215 | 75 | |
More Information
Set Membership: |
|

JADE2
Genomic Map