Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357383.3 | XP_003357431.2 | 1003 | 81 | ||
| XM_013989549.2 | XP_013845003.1 | 673 | 81 | |||
| XM_013989550.2 | XP_013845004.2 | 957 | 81 | |||
| GenBank mRNA | AK347277.1 | - | 5 - 6 | 790 | 81 | |
More Information
Set Membership: |
|

ETNK2
Genomic Map