Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074833.1 | XP_020930492.1 | 4876 | 87 | ||
| GenBank mRNA | AK347694.1 | - | 1 | 1318 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074833.1 | XP_020930492.1 | 4876 | 87 | ||
| GenBank mRNA | AK347694.1 | - | 1 | 1318 | 87 | |
Set Membership: |
|