Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986316.2 | XP_013841770.2 | 631 | 68 | ||
| GenBank mRNA | AK347711.1 | - | 3 - 4 | 639 | 68 | |
| AK393831.1 | - | 3 - 4 | 633 | 68 | ||
| AK393894.1 | - | 3 - 4 | 359 | 68 | ||
| AK400472.1 | - | 3 - 4 | 384 | 68 | ||
More Information
Set Membership: |
|

STOML2
Genomic Map