Assay Details
| Gene Symbol: TTC23L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076626.1 | XP_020932285.1 | 2474 | 81 | ||
| GenBank mRNA | AK347733.1 | - | 9 - 10 | 1282 | 81 | |
More Information
Set Membership: |
|

TTC23L
Genomic Map