Assay Details
| Gene Symbol: LYRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662093.3 | XP_005662150.1 | 558 | 62 | ||
| XM_005662094.3 | XP_005662151.1 | 555 | 62 | |||
| XM_005662096.3 | XP_005662153.1 | 430 | 62 | |||
| XM_021086518.1 | XP_020942177.1 | 376 | 62 | |||
| XM_021086519.1 | XP_020942178.1 | 268 | 62 | |||
| GenBank mRNA | AK347780.1 | - | 3 - 4 | 447 | 62 | |
| AY609502.1 | - | 3 - 4 | 357 | 62 | ||
More Information
Set Membership: |
|

LYRM1
Genomic Map