Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354739.4 | XP_003354787.1 | 825 | 60 | ||
| XM_005655210.3 | XP_005655267.1 | 850 | 60 | |||
| XM_005655211.3 | XP_005655268.1 | 848 | 60 | |||
| XM_013987876.2 | XP_013843330.1 | 822 | 60 | |||
| XM_013987877.2 | XP_013843331.1 | 778 | 60 | |||
| XM_021087295.1 | XP_020942954.1 | 681 | 60 | |||
| GenBank mRNA | AK347913.1 | - | 7 - 8 | 779 | 60 | |
More Information
Set Membership: |
|

TMEM150A
Genomic Map