Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124281.5 | XP_003124329.2 | 320 | 94 | ||
| GenBank mRNA | AK348503.1 | - | 2 - 3 | 137 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124281.5 | XP_003124329.2 | 320 | 94 | ||
| GenBank mRNA | AK348503.1 | - | 2 - 3 | 137 | 94 | |
Set Membership: |
|