Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084297.1 | XP_020939956.1 | 783 | 73 | ||
| XM_021084298.1 | XP_020939957.1 | 683 | 73 | |||
| GenBank mRNA | AK348532.1 | - | 8 - 9 | 865 | 73 | |
| AK352382.1 | - | 4 - 5 | 276 | 73 | ||
More Information
Set Membership: |
|

RNF126
Genomic Map