Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131876.4 | XP_003131924.3 | 2889 | 92 | ||
| XM_005669141.3 | XP_005669198.1 | 3775 | 92 | |||
| XM_005669142.2 | XP_005669199.1 | 3099 | 92 | |||
| GenBank mRNA | AK348673.1 | - | 1 - 2 | 99 | 92 | |
More Information
Set Membership: |
|

KIAA0753
Genomic Map