Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125437.3 | XP_003125485.1 | 248 | 96 | ||
| GenBank mRNA | AK349213.1 | - | 1 - 2 | 254 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003125437.3 transcripts.
Set Membership: |
|

LRRC24
Genomic Map