Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928562.5 | XP_001928597.3 | 513 | 83 | ||
| XM_005659178.3 | XP_005659235.2 | 524 | 83 | |||
| GenBank mRNA | AK349884.1 | - | 1 - 2 | 142 | 83 | |
| AY609677.1 | - | 1 - 2 | 130 | 83 | ||
More Information
Set Membership: |
|

HEBP2
Genomic Map