Assay Details
| Gene Symbol: MEDAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980428.2 | XP_013835882.1 | 996 | 86 | ||
| GenBank mRNA | AK350672.1 | - | 4 - 5 | 897 | 86 | |
| AY609558.1 | - | 4 - 5 | 1000 | 86 | ||
More Information
Set Membership: |
|

MEDAG
Genomic Map