Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985724.2 | XP_013841178.1 | 3416 | 93 | ||
| XM_021079001.1 | XP_020934660.1 | 3707 | 93 | |||
| GenBank mRNA | AK350842.1 | - | 1 | 925 | 93 | |
More Information
Set Membership: |
|

KBTBD2
Genomic Map