Assay Details
| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096135.1 | XP_020951794.1 | 2976 | 69 | ||
| XM_021096136.1 | XP_020951795.1 | 2778 | 69 | |||
| GenBank mRNA | AK351142.1 | - | 3 - 4 | 597 | 69 | |
More Information
Set Membership: |
|

EMILIN2
Genomic Map