Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127501.4 | XP_003127549.1 | 723 | 78 | ||
| XM_005653353.3 | XP_005653410.1 | 721 | 78 | |||
| XM_013995611.2 | XP_013851065.1 | 322 | 78 | |||
| GenBank mRNA | AK351276.1 | - | 2 - 3 | 682 | 78 | |
More Information
Set Membership: |
|

VWA1
Genomic Map