Assay Details
| Gene Symbol: CCDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127233.4 | XP_003127281.1 | 3695 | 68 | ||
| GenBank mRNA | AK352441.1 | - | 1 | 923 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127233.4 | XP_003127281.1 | 3695 | 68 | ||
| GenBank mRNA | AK352441.1 | - | 1 | 923 | 68 | |
Set Membership: |
|